Would you like to log in again?
Factor XIIIa Activity Assay Kit (Colorimetric), BioVision, Inc.
Supplier: BioVision
AB241013-100TEST
76211-886EA 1191.16 USD InStock
76211-886
Factor XIIIa Activity Assay Kit (Colorimetric), BioVision, Inc.
Assays Cellular Assays
Kit is easy-to-use and can detect Factor XIIIa (as low as 0.1 Loewy U) from plasma and purified protein samples.
  • Simple, rapid & convenient assay to measure Factor XIIIa
  • Includes Positive Control and Inhibitor (Iodoacetamide)
Factor XIII or fibrin stabilizing factor (EC 2.3.2.13) is an enzyme involved in the blood coagulation system that crosslinks fibrin. Factor XIII is a transglutaminase that circulates in the plasma as a heterotetramer of two catalytic “A” subunits and two carrier “B” subunits. Factor XIII is activated by thrombin in the presence of calcium which leads to the release of the activation peptide, followed by the dissociation of carrier subunits. The activated Factor XIII (XIIIa) acts on fibrin (Factor Ia) to form γ-glutamyl-Є-lysyl amide cross links between fibrin molecules leading to the formation of an insoluble clot. The deficiency of Factor XIII affects fibrin clot stability and bleeding disorders. BioVision's Factor XIIIa activity assay kit utilizes the transglutaminase activity of factor XIIIa to cross link an amine-containing substrate to glutamine-containing substrate resulting in the loss of ammonia which can be quantitatively measured by a colorimetric assay. The kit is easy-to-use and can detect Factor XIIIa (as low as 0.1 Loewy U) from plasma and purified protein samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty