Would you like to log in again?
UGT Activity Assay/Ligand Screening Kit (Fluorometric), BioVision
Supplier: BioVision
AB273331-100TEST
76211-892EA 960.02 USD InStock
76211-892
UGT Activity Assay/Ligand Screening Kit (Fluorometric), BioVision
Assays Cellular Assays
Simple and sensitive fluorometric assay for UGT Activity in biological samples
  • Easy-to-use
  • Non-radioactive
  • Accurately measure UGT activity in microsomes, lysates and recombinant UGT preparations
Uridine diphospho-glucuronosyltransferases (UGTs, EC 2.4.1.17) are a superfamily of microsomal membrane-bound glycosyltransferase enzymes. UGTs are responsible for the vast majority of Phase II biotransformation (conjugation) reactions, in which a hydrophilic moiety is attached to small molecule drugs and other xenobiotics to facilitate their rapid excretion from the body. UGTs are predominantly expressed in the liver, intestine and kidneys, where they catalyze the addition of a glucuronic acid moiety from the nucleotide sugar uridine-5’-diphospho-α-D-glucuronic acid (UDPGA) to nucleophilic hydroxyl, sulfhydryl or amino groups of pharmacologically active drugs, Phase I drug metabolites and certain endogenous substrates such as steroid hormones, bile acids and bilirubin. Isozymes of the UGT family have extremely broad, often overlapping substrate specificities and tend to exhibit modest affinity for substrates but high catalytic turnover rates. UGTs also contribute to ‘first-pass’ presystemic drug metabolism, which dramatically reduces the oral bioavailability of a number of drugs. BioVision’s UGT Activity Assay / Ligand Screening Kit enables rapid measurement of native or recombinant UGT activity in biological samples such as liver microsomes and can also be used to assess the effect of drugs and other novel compounds on UGT activity. The assay utilizes a highly fluorescent UGT substrate with a large Stokes shift (Ex/Em = 415/502 nm) that allows determination of UGT activity by tracking the drop in fluorescence emission as the substrate is converted into a non-fluorescent glucuronide. The multi-isozyme substrate is glucuronidated by virtually all of the pharmacologically-relevant mammalian UGT1A and UGT2B enzymes. UGT specific activity is calculated by comparing the fluorescence loss versus a control reaction performed in the absence of the required cofactor UDPGA. The kit includes the pore-forming peptide antibiotic Alamethicin, which allows the UGT Substrate and UDPGA to rapidly diffuse across lipid membranes to access the UGT active site located in the lumen of microsomes. For verification of modulation of UGT activity by test ligands, diclofenac, a competitive inhibitor of most human and rodent UGT isozymes, is also included. The assay is highly sensitive, simple to perform and high-throughput adaptable. This assay can detect less than 0.1 mU UGT activity in biological samples. The kit contains a complete set of reagents sufficient for performing 100 reactions at a 100 μl reaction volume
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty