Would you like to log in again?
Anoikis Assay kit, BioVision, Inc.
Supplier: BioVision
AB284938-100TEST
76211-896EA 1078.11 USD InStock
76211-896
Anoikis Assay kit, BioVision, Inc.
Assays Cellular Assays
Our assay is easy to use, sensitive and compatible with fluorimeters
  • Highly sensitive fluorometric method to measure cell Anoikis in response to a variety of biochemical stimuli
  • Simple & High throughput-adaptable
  • Reproducible, Quantitative tool for screening, studying, and characterizing compounds that affect Anoikis
Anoikis is a form of cellular apoptosis induced by the inability of normal cells to attach to the extracellular matrix (ECM). Incorrect attachment or no ECM attachment could affect cell growth and differentiation in normal cells. However, research studies has determined that metastatic cells can undergo Anoikis without affecting their ability to migrate and invade surrounding or distant organs. BioVision’s Anoikis assay kit quantifies live cells using fluorometric (Calcein AM) reagents and a proprietary coated chamber that mimics Anoikis conditions. Our assay is easy to use, sensitive and compatible with fluorimeters
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty