Would you like to log in again?
Phospho-ERK1/2 (Thr202/Tyr204) Translocation Assay Kit (Cell-Based), BioVision, Inc.
Supplier: BioVision
AB284557-50TEST
76211-846EA 1049.53 USD InStock
76211-846
Phospho-ERK1/2 (Thr202/Tyr204) Translocation Assay Kit (Cell-Based), BioVision, Inc.
Assays Cellular Assays
A cell-based assay kit that detect the nuclear translocation of activated ERK1/2 in fixed mammalian cells.
  • Detection of subcellular localization of activated ERK1/2 in mammalian cells
  • Screening and characterizing effectors of ERK1/2 kinases
  • Studying of cell signaling, cell division and cell proliferation mechanism
Mitogen-activated protein kinases (MAPKs) are proline-directed serine and threonine protein kinases that regulate numerous physiological cell responses including: embryogenesis, cell differentiation, proliferation, migration, apoptosis and death. Extracellular signal-regulated kinases (ERKs) 1 and 2 (ERK1/2), also known as p44 MAPK and p42 MAPK respectively, belong to one of the five major groups of MAPKs. Closely-related ERK1/2 isoforms are uniquely activated by several extracellular signals including growth factors, cytokines, hormones, and neuro-transmitters. Activation of ERK1/2 by the upstream kinases MEK1 and MEK2 occurs via dual phosphorylation on specific threonine (Thr202) and tyrosine (Tyr204) residues on the T*EY* motif. MEK1 and MEK2 are activated through receptors (tyrosine kinases or integrins) via pathways involving adaptor proteins, guanine nucleotide exchange factors, small GTP binding proteins, and MAPKKs. Activated ERK1/2 phosphorylates both, cytosolic (SOS, MNK1/2, RSKs) and nuclear targets. In the nucleus, it affects gene expression and DNA replication by the phosphorylation of MSK 1 and 2 and the transcription factors Elk-1, Sap1, and Sap2. In cultured cells, growth factors or mitogens induce rapid and transient translocation of activated ERK1/2 to nucleus. Different cell lines exhibit various duration, magnitude, and subcellular localization of activated/phosphorylated ERK1/2. The response of the protein may differ even within the same cell line depending on the dose and cell density. BioVision’s Phospho-ERK1/2 (Thr202/Tyr204) Translocation Assay Kit provides a simple and complete assay in a ready-to-use format to visualize the translocation of activated ERK1/2 between cytoplasmic and nuclear compartments in mammalian cells.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty