Would you like to log in again?
Angiotensin II Converting Enzyme (ACE2) Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB273297-100TEST
76211-898EA 921.97 USD InStock
76211-898
Angiotensin II Converting Enzyme (ACE2) Activity Assay Kit (Fluorometric), BioVision
Assays Enzyme Assays
Highly Sensitive; Includes human ACE2 positive control
  • Simple one-step reaction
  • Takes only 1-2 hrs
  • Non-radiometric fluorescent detection
  • HTP adaptable
Angiotensin II converting enzyme (ACE2, EC 3.4.17.23), a zinc-based metalloprotease, is part of the renin-angiotensin system (RAS) that controls the regulation of blood pressure by cleaving the C-terminal dipeptide of Angiotensin II to convert it into Angiotensin 1-7. ACE2 is a receptor of human coronaviruses, such as SARS and HCoV-NL63. It is expressed on the vascular endothelial cells of lung, kidney and heart. ACE2 is a potential therapeutic target for cardiovascular and coronavirus-induced diseases. BioVision’s ACE2 Activity Assay Kit kit will help the research progress in this field. This kit utilizes the ability of an active ACE2 to cleave a synthetic MCA based peptide substrate to release a free fluorophore. The released MCA can be easily quantified using a fluorescence microplate reader. We also provide an ACE2 specific inhibitor that can differentiate the ACE2 activity from other proteolytic activity.The kit can detect as low as 0.4 mU. Our assay kit is simple and can be used in a high-throughput format
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty