Would you like to log in again?
Cell Transformation Assay Kit (Colorimetric), BioVision
Supplier: BioVision
AB235698-100TEST
76211-900EA 1095.53 USD InStock
76211-900
Cell Transformation Assay Kit (Colorimetric), BioVision
Assays Cellular Assays
Highly Sensitive and Quantitative (Colorimetric), HTP adaptable Assay to measure Cell Transformation
  • Highly sensitive fluorometric method to measure cell transformation in response to a variety of biochemical stimuli
  • Simple & High throughput-adaptable
  • Reproducible, Quantitative tool for screening, studying, and characterizing compounds that affect cell transformation
The transformed cells can proliferate without attaching to surface. Anchorage independent cell growth is the hallmark of cell transformation. The Soft-Agar Assay is a traditional method for screening cell transformation in vitro. However, this method is lengthy (3-4 weeks incubation), laborious (counting colonies) and inconsistent (due to subjective counting). BioVision’s Cell Transformation Assay is faster, stable, and more sensitive than the traditional soft-agar assay. The kit is based on the conversion of the tetrazolium salt (WST-8) to formazan by cellular mitochondrial dehydrogenases. The generated signal is directly proportional to the number of living cells. This one-step method is non-radioactive and simple (just add-and-read, does not require harvesting cells, and solubilization steps). The assay is high-throughput adaptable and has wide linear range from 10000-400000 cells. The entire Cell Transformation Assay can be finished within 7-8 days.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty