Would you like to log in again?
Cell Transformation Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB235699-100TEST
76211-902EA 1208.56 USD InStock
76211-902
Cell Transformation Assay Kit (Fluorometric), BioVision
Assays Cellular Assays
Highly Sensitive and Quantitative (Fluorometric), HTP adaptable Assay to measure Cell Transformation
  • Highly sensitive fluorometric method to measure cell transformation in response to a variety of biochemical stimuli
  • Simple & High throughput-adaptable
  • Reproducible, Quantitative tool for screening, studying, and characterizing compounds that affect cell transformation
Transformed cells can proliferate without attaching to surface Anchorage-independent cell growth is the hallmark of cell transformation. The Soft-Agar Assay is a traditional method for screening cell transformation in vitro. However, this method is lengthy (3-4 weeks incubation), laborious (counting colonies) and inconsistent (due to subjective counting). BioVision’s Cell Transformation Assay is faster, stable, and more sensitive than the traditional soft-agar assay. The kit uses a quantitative dye that binds to nucleic acid and generates green fluorescence. This one-step method is non-radioactive and simple (just add-and-read, and does not require tedious labor such as counting colonies). The assay is high-throughput adaptable and has wide linear range from 50-60,000 cells. The entire assay can be finished within 7-8 days.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty