Would you like to log in again?
Cardiolipin Assay Kit (Fluorometric), BioVision, Inc.
Supplier: BioVision
AB241036-100TEST
76211-850EA 1141.51 USD InStock
76211-850
Cardiolipin Assay Kit (Fluorometric), BioVision, Inc.
Assays Cellular Assays
The easiest kit in the market to measure Cardiolipin in samples
  • Simple, fast, does not require lenghty incubation times
Cardiolipin (CL) is a very important phospholipid present in the inner mitochondrial membrane and constitutes about 20% of its total lipid composition. It has a dimeric structure comprised of two phosphatidic acid residues linked by a glycerol bridge. It is essential for several mitochondrial functions such as maintaining activity of the electron transport chain complexes, and other mitochondrial processes including biogenesis, fission, fusion and protein transport. It is involved in apoptosis where it helps alter the mitochondrial membrane structure and aids in the release of cytochrome c. Condition such as diabetes and heart failure are linked to changes in levels of cardiolipin and it is also depleted during aging. Exposure to toxicants like cigarette smoke and organophosphates may cause an alteration in cardiolipin levels and composition, thus adversely affecting health. BioVision’s Cardiolipin Assay is a fluorometric assay that makes use of a proprietary probe that fluoresces on association with cardiolipin but not with any other lipids such as phosphatidylcholine and sphingomyelin, making it highly specific. Fluorescence can be recorded at Ex/Em 340/480 nm. The kit includes purified cardiolipin as standard and can detect as low as 0.2 nmol of cardiolipin
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty