Would you like to log in again?
3D Cell Culture HTS Cell Viability Complete Assay Kit, BioVision, Inc.
Supplier: BioVision
AB284562-100TEST
76211-906EA 660.55 USD InStock
76211-906
3D Cell Culture HTS Cell Viability Complete Assay Kit, BioVision, Inc.
Assays Cellular Assays
Kit provides an easy, non-radioactive, high-throughput method for screening cytotoxicity & apoptosis
  • Convenient & non-radioactive
  • High-throughput
  • Highly-sensitive: detect as low as 50 viable cells in less than 30 min.
  • Suitable for cell proliferation, cell viability, chemotaxis, cytotoxicity and apoptosis
Three dimensional (3D) cell cultures are artificially-created environments in which cells are permitted to grow or interact with their surroundings in a 3D fashion. 3D cell cultures improve the function, differentiation and viability of cells and recapitulate in vivo microenvironment compared to conventional 2D cell cultures. 3D matrices provide a physiologically relevant screening platform, by mimicking the in vivo responses, for many cell types including cancer and stem cells in developmental morphogenesis, pharmacology, drug metabolism and drug toxicity studies. Quantification of number of viable cells is an indispensable tool in in vitro screening in these studies. Calcein AM is a non-fluorescent, hydrophobic compound that easily penetrates intact and live cells, and has been widely used to assess cell viability and proliferation in Cell Biology research. However, with the use of 3D matrices, some proteases-based dissociation methods don’t completely dissolve the matrices and cell aggregates, which could alter the result in quantitative in vitro assays such as viability assessment. BioVision’s 3D Cell Culture Cell Viability Complete Assay Kit provides a standardized fluorometric method for sensitive quantification of viable cells that can detect as low as 50 viable cells in each well and can be measured at Ex/Em = 485/530 nm. The measured fluorescence intensity is proportional to the number of viable cells. Further, as a complete set, the kit comes with an optimized and gentle non-enzymatic dissociation solution for the recovery of viable and dead cells from spheroids in matrices and scaffolds. This assay kit provides an easy-to-use, non-radioactive, and high-throughput method for characterizing and screening cell viability, cytotoxicity and apoptosis.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty