Would you like to log in again?
EZScreen™ Triglyceride Quantification Colorimetric Assay Kit, BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB282924-400TEST
76211-910EA 1083.34 USD InStock
76211-910
EZScreen™ Triglyceride Quantification Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Highly Sensitive, no sample preparation required
  • Simple procedure; takes ~40 minutes
  • Fast and convenient
  • Kit provides all necessary buffers and reagents for a accurate assay of triglyceride in various mammalian samples
Triglycerides (TGs) are the main constituent of body fat in humans and animals, as well as vegetable fat. TGs are found as a natural component in human’s blood circulatory system. TGs are composed of one molecule of glycerol and 3 molecules of fatty acids. The metabolism of cholesterol generates fatty acids and glycerol, and both can serve as substrates in metabolic pathways to produce energy. Excess TGs can remain in the body as fatty deposits and that may lead to obesity. Elevated triglyceride levels also increase the risk for heart disease, strokes, and type 2 diabetes.  BioVision’s EZScreenTM Triglyceride Quantification Kit provides a sensitive, easy assay to measure TG concentration in a variety of samples. In the assay, TGs are converted to free fatty acids and glycerol. The released glycerol is then oxidized to generate a product which reacts with a probe to generate a color that can be detected by a spectrophotometer at λ = 590 nm. The method is quantitative, rapid, simple, sensitive, and designed for high throughput format. The kit provides a convenient means for detecting 0.5 to 5 mM Triglycerides in biological samples
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty