Would you like to log in again?
EZScreen™ Beta-Lactamase Activity Colorimetric Assay Kit, BioVision
Supplier: BioVision
AB282926-400TEST
76211-596EA 1478.08 USD InStock
76211-596
EZScreen™ Beta-Lactamase Activity Colorimetric Assay Kit, BioVision
Assays Enzyme Assays
Highly Sensitive, no sample preparation required
  • Rapid, simple & sensitive
  • The assay can detect enzymatic activity as low as 15 mU in a variety of biological samples
Beta-Lactamases (βLs), are a large family of hydrolases comprising more than 850 identified members expressed in Gram-positive and Gram-negative bacteria. βLs can be classified according to their substrate or inhibitor specificity. These enzymes are capable of hydrolyzing four atom rings known as β-lactams. Antibiotics containing β-lactam rings (i.e. penicillin, cephalosporin, monobactam, carbapenem) are highly susceptible to be hydrolyzed via enzymatic activity, which deactivates their antibiotic potency. βLs have become a significant clinical threat due to the alarming number of cases of bacterial strains showing β-lactam antibiotic resistance. Approaches for combatting this type of resistance have opened up the possibility of developing a new form of β lactam antibiotics that are more resistant to β-Lactamase enzymatic activity. Therefore, experimental research has been arising in wide extent for developing new drugs or inhibitors effective against the β-lactam antibiotic resistance bacteria. BioVision’s EZScreenTM Beta-Lactamase Activity Assay Kit offers a simple and sensitive assay that can detect and quantify the enzymatic activity of these hydrolases. The assay is based on the hydrolysis of Nitrocefin, a chromogenic cephalosporin, that results in the generation of a colored product (OD: 490 nm), which is directly proportional to the amount of βL activity. The assay can detect enzymatic activity as low as 15 mU in a variety of biological samples
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty