Would you like to log in again?
EZScreen™ ATP Colorimetric Assay Kit, BioVision
Supplier: BioVision
AB282930-400TEST
76211-916EA 1234.64 USD InStock
76211-916
EZScreen™ ATP Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Highly Sensitive, minimum sample preparation required
  • Simple procedure; takes less than 1 hour
  • Fast and convenient
  • Kit is designed to be a robust method which enzymatic detection of ATP in numerous biological samples
Adenosine Triphosphate (ATP) is a derivative of adenosine nucleotide that contains a large amount of biochemical energy stored in its high-energy phosphate bonds. ATP releases energy when it is hydrolyzed into ADP or AMP (Adenosine Di or Mono Phosphate). This energy is used by cells to drive various cellular events like metabolic processes, enzymatic reactions, motility, cell division and muscle contraction. It is also used as a substrate in signal transduction, by kinases to phosphorylate proteins and lipids, and by adenylate cyclase to produce cyclic AMP. In eukaryotic cells, ATP is produced via cellular respiration in mitochondria and by photosynthesis in chloroplasts. Anaerobic bacteria can produce ATP by breaking down nutrients into simpler metabolites, and then using the released energy to form ATP from ADP and Pi (inorganic phosphate). Aerobic bacteria also make ATP from ADP and Pi by ATPase which takes the advantage of ‘proton motive force’ when protons travel back to cytosol from periplasmic space. There are many commercially available kits that detects ATP in femtomoles or less by using luminescence but these products require specialized instrumentation and utilize luciferase which can be difficult to maintain in active form. BioVision’s EZScreenTM ATP Colorimetric Assay Kit is designed to be a robust, simple, stable, colorimetric method which utilizes a series of enzymatic reactions to form a product that is easily quantified at OD 570 nm. The method is rapid, simple, sensitive and designed for high-throughput format using a 384-well plate. The kit can detect as low as 80 pmol of ATP in a 384 well assay plate.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty