Would you like to log in again?
p53 Nuclear Translocation Assay Kit (Cell-Based), BioVision
Supplier: BioVision
AB284563-50TEST
76211-920EA 1156.78 USD InStock
76211-920
p53 Nuclear Translocation Assay Kit (Cell-Based), BioVision
Assays Cellular Assays
Easy & complete assay kit to visualize the activation & nuclear translocation of p53 in human cells.
  • Simple procedure to visualize p53 nuclear translocation in ~ 3 hours
  • Fast and contains Nutlin-3 that serves as a control to induce p53 translocation from the cytoplasm to the nucleus.
p53 is a tumor suppressor gene which encodes a transcription factor that controls cell’s destiny. This gene gets activated when cells are exposed to increased cellular stress, or DNA damage. The levels of p53 are elevated when post-translational modifications occur, which block its sequestration and/or ubiquitination by its destabilizer, Mouse Double Minute 2 Homolog (MDM2) which in turn is a transcriptional target of p53. Due to the three putative nuclear localization signals (NLSs) on its C-terminus, some of the activated p53 protein translocates into the nucleus and activate genes that induce cell cycle arrest, senescence, or apoptosis. The p53 gene is frequently mutated in cancer cells, and about 50% of cancers show p53 mutations, resulting in loss of its functions. The loss of p53 functionality can lead to dysregulation of many biological processes in cells, such as metabolic pathways, cellular homeostasis, cell movement, apoptosis, etc. BioVision’s p53 Nuclear Translocation Assay Kit (Cell-Based) provides an easy and complete assay kit to visualize the activation and nuclear translocation of p53 in human cells. This assay kit uses BioVision’s specific and sensitive human p53 antibody and a p53 secondary antibody to visualize the localization of p53 in fixed common human cells along with DAPI, a fluorescent stain, for nuclear staining. The kit includes Nutlin-3, a potent selective inhibitor that disrupts the protein-protein interaction between p53 and MDM2. Nutlin-3 serves as a control to induce p53 translocation from the cytoplasm to the nucleus
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty