Would you like to log in again?
Prostate Specific Antigen (PSA) Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB273302-100TEST
76211-922EA 873.34 USD InStock
76211-922
Prostate Specific Antigen (PSA) Activity Assay Kit (Fluorometric), BioVision
Assays Enzyme Assays
Highly Sensitive, minimum sample preparation required
  • Simple one-step reaction
  • Takes only 1-2 hrs
  • Non-radiometric fluorescent detection
  • HTP adaptable
Prostate Specific Antigen (PSA, EC 3.4.21.77), or Kallikrein-related peptidase 3, is a serine protease. It proteolyses the Semenogelin I and Semenogelin II of seminal fluid and thus liquefies it. PSA also shows antiangiogenic activity. The significance of the level of enzymatically active PSA is emerging in diagnosis and prognosis of prostate cancer. Growing number of evidence suggests that PSA plays an important role in prostate tumor growth, invasion and metastasis. BioVision’s PSA Activity Assay Kit enables the research in this field. This kit utilizes the ability of an active PSA to cleave a synthetic AMC based peptide substrate to release a free fluorophore. The released AMC can be easily quantified using a fluorescent microplate reader. This assay kit is simple and can be used in a high-throughput format. This assay can detect as low as 1 ng of purified active PSA.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty