Would you like to log in again?
mCherry Quantification Kit, BioVision
Supplier: BioVision
AB284566-100TEST
76211-932EA 643.42 USD InStock
76211-932
mCherry Quantification Kit, BioVision
Assays Cellular Assays
Kit provides an easy 96 micro-plate assay to analyze mCherry expression level in cells or tissues
  • Simple procedure; takes ~ 20 minutes
  • Fast and convenient
mCherry is the second generation monomeric red fluorescent protein that was derived from proteins originally isolated from Cnidarians (jelly fish, sea anemones and corals), such as GFP or DsRed. mCherry is widely used as a fluorescent tracer in transfection and transgenic experiments because of its better performance in brightness, photostability and monomeric structure. Since mCherry requires no additional substrates or cofactors, mCherry can be easily detected under a fluorescence microscope. However, most imaging studies of mCherry are only qualitative. BioVision’s mCherry Quantification Kit provides an easy 96 micro-plate assay to analyze mCherry expression level in cells or tissues quantitatively. Cells or tissues can be homogenized directly in the mCherry Assay Buffer. The amount of mCherry is determined by comparing its fluorescence (Ex/Em= 587/610 nm) with a mCherry standard that is provided in this kit. Each kit provides sufficient reagents to perform up to 100 assays.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty