Would you like to log in again?
Factor XIa Activity Assay Kit (Colorimetric), BioVision, Inc.
Supplier: BioVision
AB241037-100TEST
76211-936EA 1191.16 USD InStock
76211-936
Factor XIa Activity Assay Kit (Colorimetric), BioVision, Inc.
Assays Cellular Assays
Highly Sensitive, minimum sample preparation required
  • Simple, rapid & convenient assay to measure Factor XIa
  • Includes Positive Control and Factor Xia and Factor XIIa Inhibitors
Factor XI or plasma thromboplastin antecedent (EC 3.4.21.27), the zymogen form of factor XIa, is a serine protease. In humans, Factor XI is encoded by the F11 gene. Factor XI (FXI) circulates as a homo-dimer in its inactive form activated into factor XIa by factor XIIa (FXIIa) via the "contact pathway", thrombin, and self-activation by its active form (FXIa). The deficiency of factor XI causes the rare hemophilia C which is an autosomal recessive disorder characterized by lower spontaneous bleeding, but excessive blood loss during surgical procedures. Low levels of factor XI also occur in many other disease states, including Noonan syndrome. High levels of factor XI have been implicated in thrombosis. Factor XIa also activates factor IX which, in turn, activates factor X in the coagulation cascade. BioVision's Factor XIa activity assay kit utilizes the ability of factor XIa to cleave a synthetic substrate to release p-Nitroaniline (pNA) which can be quantitatively measured by a colorimetric assay (OD 405 nm). The kit is easy-to-use and can detect Factor XIa (as low as 1 mPEU) from plasma and purified protein samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty