Would you like to log in again?
Glycerol Cell-Based Assay Kit, BioVision, Inc.
Supplier: BioVision
AB235701-100TEST
76211-942EA 856.72 USD InStock
76211-942
Glycerol Cell-Based Assay Kit, BioVision, Inc.
Assays Cellular Assays
Highly Sensitive Assay. Cell-based assay to detect total glycerol in cultured cells.
  • Simple procedure; takes ~ 2 hours
  • Fast and convenient
  • The kit provides an accurate assay for measuring free glycerol in cell-based assays
Metabolic diseases such as type II diabetes and fatty liver are becoming more prevalent among an aging population, and are a burden to the healthcare system. Such diseases revolve around the body’s ability to control triglyceride (lipid) metabolism. Events such as fasting can lead to cell autophagy, corresponding with removal of lipid droplets. A number of pathologies can lead to steatosis, bringing about triacylglyceride buildup and release of free fatty acids and free glycerol. Glycerol molecules can then be reabsorbed by the liver or kidney and can be metabolized to re-enter the energy store by pathways such as glycolysis and gluconeogenesis. The glycerol concentration can thus be used as a measure of lipid metabolism and fat mobilization. BioVision’s Glycerol Cell-Based Assay Kit provides a simple, sensitive, straight-forward assay for determination of glycerol concentrations in various cell and tissue culture samples. The assay uses a glycerol-specific enzymatic reaction to convert the probe into a fluorescent product in the presence of glycerol. Glycerol levels can be detected as low as 50 pmol per well, or 1.0 µM in culture medium. Chloroquine (included in the kit as a control) induces steatosis and blocks autophagy, thus leading to a buildup of lipid droplets and an increase in the free glycerol concentration.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty