Would you like to log in again?
Nitric Oxide Cell-Based HTS Assay Kit, BioVision
Supplier: BioVision
AB284567-100TEST
76211-946EA 987.78 USD InStock
76211-946
Nitric Oxide Cell-Based HTS Assay Kit, BioVision
Assays Cellular Assays
Ultrasensitive, Simple, HTP adaptable fluorometric assay kit for measuring NO concentration
  • Simple, HTP adaptable protocol
  • Sensitivity as low as 5 pmol of nitric oxide in cells
  • Fluorometric detection
Nitric oxide (NO) plays an important role in neurotransmission, vascular regulation, immune response and apoptosis. NO has been established as one of the key regulators in cardiovascular, nervous, and immune systems. NO plays a pivotal role in numerous processes: in central nervous system, NO participates in cell communication and information storage; in vascular endothelium, NO is involved with regulation of vascular function. Finally, NO is produced by immune cells, including macrophages, as a part of body’s defense mechanism during immunological responses and oxidative stress conditions. However, chronic overproduction of NO is one of the fundamental causes underlying disorders including neurodegenerative diseases and pathophysiology of blood vessels. BioVision’s Nitric Oxide Cell-Based HTS Assay Kit utilizes a dye that reacts with intracellular NO to produce a highly-Fluorescent Triazole Product (Ex/Em= 488/532 nm). The kit includes Diphenyleneiodonium (DPI), a potent inhibitor of nitric oxide cellular production, which has been widely used to inhibit NO synthesis in order to evaluate NO function in different systems and diseases. This kit provides a simple, standardized and HTS method to quantitate the amount of NO in cell-based assays. This Assay Kit is user-friendly, sensitive and can detect the NO Fluorescent product as low as 5 pmol in a 96-well plate
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty