Would you like to log in again?
5-Lipoxygenase Inhibitor Screening Kit (Fluorometric), BioVision, Inc.
Supplier: BioVision
AB284521-100TEST
76211-948EA 1112.9 USD InStock
76211-948
5-Lipoxygenase Inhibitor Screening Kit (Fluorometric), BioVision, Inc.
Assays Enzyme Assays
Highly Sensitive; Includes recombinant Zileuton as inhibitor control
  • Simple procedure; takes only ~40 min
  • Fast and convenient
5-Lipoxygenase (5-LOX) is a non-heme iron-containing dioxygenase that converts unsaturated fatty acids to epoxides; for example the synthesis of leukotrienes from arachidonic acid. It is involved in processes like cell proliferation, differentiation and inflammation and has been implicated in inflammation and hyper proliferation mediated diseases like asthma, rheumatoid arthritis and cancer. Research for identifying potential remedies for such diseases involves the identification of inhibitors of 5-lipoxygenase. BioVision’s 5-Lipoxygenase Inhibitor Screening Kit uses Zileuton, a well-known 5-lipoxygenase inhibitor as positive control, and provides a simple and fast method to screen/ characterize potential 5-lipoxygenase inhibitors
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty