Would you like to log in again?
Plasma Kallikrein Inhibitor Screening Kit (Colorimetric), BioVision
Supplier: BioVision
AB284523-100TEST
76211-966EA 1109.75 USD InStock
76211-966
Plasma Kallikrein Inhibitor Screening Kit (Colorimetric), BioVision
Assays Cellular Assays
High-throughput adaptable assay kit can be used to screen potential inhibitors of Plasma Kallikrein.
  • Simple and reliable test to screen Plasma Kallikdrein inhibitors/activators
  • High-throughput compatible
  • Includes Inhibitor Control
Plasma Prekallikrein (EC 3.4.21.34), is the glycosylated single chain zymogen precursor of the plasma serine protease Kallikrein (PK). It circulates with kininogen and is activated by Factor XIIa to Kallikrein in the intrinsic coagulation pathway. Kallikrein activates plasminogen in fibrinolysis and cleaves kininogen in the bradykinin system of vasodilation. Prekallikrein deficiency is rare and causes increased activated partial thromboplastin time. Elevated plasma Prekallikrein is associated with diabetes and cardiovascular disease. Plasma Kallikrein inhibitors have been proposed as drugs to manage Hereditary Angioedema. BioVision’s Plasma Kallikrein Inhibitor Screening Kit utilizes the ability of active Plasma Kallikrein to cleave a synthetic pNA-based peptide substrate to release pNA (OD405 nm), which can be easily quantified using a microplate reader. In the presence of a Plasma Kallikrein inhibitor, the cleavage of this substrate is reduced/abolished resulting in decrease or total loss of the pNA absorbance. This simple and high-throughput adaptable assay kit can be used to screen/study/characterize potential inhibitors of Plasma Kallikrein.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty