Would you like to log in again?
Senescence Detection Kit (Fluorometric), BioVision, Inc.
Supplier: BioVision
AB228562-100TEST
76211-372EA 1356.34 USD InStock
76211-372
Senescence Detection Kit (Fluorometric), BioVision, Inc.
Assays Cellular Assays
Simple, sensitive, one-step assay to measure senescence cells using FACS
  • Simple, one step protocol
Senescence is thought to be a tumor suppressive mechanism and an underlying cause of aging. Senescence represents an arrested state in which the cells remain viable, but not stimulated to divide by serum or passage in culture. Senescent cells display increase of cell size, senescence-associated expression of β-galactosidase (SA-β-Gal) activity, and altered patterns of gene expression. The Senescence detection kit is designed to fluorescently detect SA-β-Gal activity in cultured cells by FACS. The SA-β-Gal is present only in senescent cells and is not found in pre-senescent, quiescent or immortal cells
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty