Would you like to log in again?
5’-Nucleotidase Activity Assay Kit (Colorimetric), BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB235945-100TEST
76211-374EA 1034.21 USD InStock
76211-374
5’-Nucleotidase Activity Assay Kit (Colorimetric), BioVision
Assays Enzyme Assays
Simple, sensitive, enzymatic assay to measure 5'NT Activity in biological samples
  • Simple, highly sensitive, high-throughput compatible
5’-Nucleotidase (5’-NT), also known at CD73 (EC 3.1.3.5) is an enzyme located in the plasma membrane. It converts extracellular nucleotides like 5’-AMP to their corresponding nucleosides, through phosphorylitic cleavage. This conversion facilitates uptake of the nucleosides through nucleoside receptors into the cell, where they can again be phosphorylated to generate nucleotides and contribute to the nucleotide pool, inside the cell. 5’-NT levels are elevated in hepatic diseases such as viral hepatitis, alcoholic liver disease and cirrhosis. BioVision’s 5’-Nucleotidase Activity Kit is a simple two-step end point assay that relies on the Berthelot’s test for quantification of ammonia. In this assay, the action of 5’-nucleotidase on the substrate generates a product, which releases ammonia in presence of the converter. Developer I and II are then used to quantify the released ammonia through increase in absorbance at 670 nm. This assay can detect as low as 0.2 mU of 5’-NT. Since non-specific enzymes like alkaline phosphatase can give a positive signal in this assay, 5’-NT inhibitor may be used to completely inhibit 5’-nucleotidase and distinguish from the signal from non-specific enzymes. The assay kit also includes 5’- Nucleotidase (5’-NT) enzyme for use as positive control
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty