Would you like to log in again?
Factor XIIa Activity Assay Kit (Colorimetric), BioVision, Inc.
Supplier: BioVision
AB241041-100TEST
76211-376EA 1049.54 USD InStock
76211-376
Factor XIIa Activity Assay Kit (Colorimetric), BioVision, Inc.
Assays Cellular Assays
Highly Sensitive, minimum sample preparation required
  • Simple, rapid & convenient assay to measure Factor XIa
  • Includes Positive Control and Factor XIIa and Factor XIIa Inhibitors
Factor XII or Hageman factor (EC 3.4.21.38), is the zymogen form of factor XIIa, a serine protease involved in Coagulation Pathway. This single chain zymogen is activated by Kallikrein into a two-chain serine protease (XIIa) with a heavy chain (factor XIIa-alpha) and a light chain. Human Factor XII and Prekallikrein are thought to be involved in a reciprocal activation mechanism in which Factor XIIa activates Prekallikrein to Kallikrein, which in turn converts Factor XII to XIIa. Factor XIIa activates Factor XI to XIa thereby triggering the Contact Factor cascade. The defects in this gene do not cause any clinical symptoms but prolong the whole blood clotting time. BioVision's Factor XIIa activity assay kit utilizes the ability of factor XIIa to cleave a synthetic substrate to release p-Nitroaniline (pNA) which can be quantitatively measured by a colorimetric assay (OD405 nm). The kit is easy-to-use and can detect Factor XIIa (as low as 1 mPEU) from plasma sampless.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty