Would you like to log in again?
Neprilysin Inhibitor Screening Kit (Fluorometric), BioVision
Supplier: BioVision
AB284525-100TEST
76211-604EA 1269.41 USD InStock
76211-604
Neprilysin Inhibitor Screening Kit (Fluorometric), BioVision
Assays Cellular Assays
High-throughput compatible, quick and easy screening for Neprilysin inhibitors.
  • Simple and reliable test to screen Neprilysin inhibitors/activators
  • High-throughput compatible
  • Includes Inhibitor Control (Thiorpan)
Neprilysin (NEP, EC 3.4.24.11), also known as neutral endopeptidase, enkephalinase, CD10, and common acute lymphoblastic leukemia antigen, is a type II zinc-containing transmembrane metalloproteinase. It is able to hydrolyze very important endogenous peptides, such as natriuretic atrial factor, enkephalins, substance P, bradykinin and amyloid beta-peptide. Thus, NEP is a potentially therapeutic target in important pathological conditions such as cardiovascular disease, prostate cancer, Alzheimer’s disease and analgesia. NEP has also been used as a biological marker of a type of childhood leukemia called CALLA. NEP is currently a focus of interest in cardiovascular and neurobiological research. NEP inhibitors have shown promising results as potential treatment alternative for some conditions including hypertension, heart failure, renal failure, and improvement of β-cell function in obese type 2 diabetes mellitus. The combination of NEP inhibitor and angiotensin II type 1 receptor blockade has been used clinically to treat patients with chronic heart failure. BioVision’s NEP Inhibitor Screening Kit can be used to screen for potential inhibitors of NEP activity. It utilizes the ability of an active NEP to cleave a synthetic MCA based peptide substrate to release a free fluorophore. The released MCA can be easily quantified using a fluorescence microplate reader. In the presence of an NEP specific inhibitor, the enzyme loses its activity which results in decrease of fluorescence intensity. The assay kit is simple and can be used to identify and characterize NEP inhibitors in a high-throughput format
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty