Would you like to log in again?
Cystathionine β Synthase Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB241043-100TEST
76211-608EA 1147.69 USD InStock
76211-608
Cystathionine β Synthase Activity Assay Kit (Fluorometric), BioVision
Assays Enzyme Assays
First assay in the market that measures CβS activity in biological samples
  • Simple, convenient, highly sensitive
  • Fluorescent assay enables easy determination of CβS activity in a variety of biological samples
  • Kit includes a stable CβS positive control
Cystathionine β Synthase (EC 4.2.1.22, CβS) is a PLP-dependent enzyme that catalyzes the formation of H2S and cystathionine when using cysteine and homocysteine as substrates. Known for its role in human sulfur metabolism, mutations in the gene encoding CβS can result in high concentrations of homocysteine in plasma. Elevated circulating concentrations of homocysteine result in genetic disorders including Homocysteinuria and Down syndrome. Specifically, Homocysteinuria is an autosomal recessive disease with clinical manifestations including mental retardation, thromboembolism and connective tissue defects. Hydrogen sulfide, CβS product, is an important gaseous mediator, like nitric oxide, that has significant effects on the immunological, neurological, cardiovascular and pulmonary systems of mammals. BioVision’s Cystathione β Synthase Assay Kit utilizes cysteine and homocysteine as substrates to produce H2S. Hydrogen sulfide reacts with the azido-functional group of the fluorescent probe yielding a fluorescent amino group (Ex/Em = 368/460 nm). The assay is highly sensitive, has a simple easy-to-follow protocol, and can detect as low as 1.45 mU of CβS activity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty