Would you like to log in again?
Metal Oxidation Catalyst Kit, PhosphonicS
Supplier: STREM CHEMICALS INC MS
96-6770-1KIT
101419-184EA 935.57 USD InStock
101419-184
Metal Oxidation Catalyst Kit, PhosphonicS
Metal Oxidation Catalyst Kit
This Kit Contains 5g each of the following Strem Product #s:
58-5100 Cerium(IV) ethyl/butyl phosphonate Silica (PhosphonicS POCe)
27-0900 Cobalt(II) ethyl/butyl phosphonate Silica (PhosphonicS POCo)
25-1200 Manganese(II) ethyl/butyl phosphonate Silica (PhosphonicS POMn)
23-4380 Vanadyl(II) ethyl/butyl phosphonate Silica (PhosphonicS POVO)
Please check strem.com for any updates and changes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty