Would you like to log in again?
Top Dog 6 Mil Sleeves, 18" Length, Remco Products
Supplier: Remco
Top Dog
84012 84014 84015 84013 84016
76196-532CS 308.03 USD InStock
76196-532 76196-530 76196-528 76196-526 76196-534
Top Dog 6 Mil Sleeves, 18" Length, Remco Products
Workwear
Reusable and washable, the one-piece construction increases the sleeve's hygiene and durability.
  • Resistance to chemical and acid sanitation products
  • Non-porous surface will not host bacteria or mold
  • Washer and dryer safe
  • Will not crack or stiffen in low temperatures or with repeated cleaning
  • Impervious to animal fats and degreasers
  • Completely latex-free for sensitive users
  • Easily repairable
These sleeves are made of 6 mil pure polyurethane. They provide coverage from above the elbow to the wrist, with elastic at both ends to keep them in place. Five colors make the sleeves coordinate with Top Dog's aprons and Remco and Vikan's standard product line colors. The sleeves add extra coverage while providing worker protection and flexibility, both in design and in function.
Certifications: Made of FDA Compliant material.
Ordering information: Sleeves can only be purchased by the case. Case quantity is 24.
Delivery information: Dropshipping available.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty