Would you like to log in again?
Vacuum Trap, Chemglass
Supplier: Chemglass
CG-4530-01
80072-656EA 835.56 USD InStock
80072-656
Vacuum Trap, Chemglass
Vacuum Traps
Trap is designed for high volume solvent removal by vacuum.
The unique design of the stopper decreases the chance of freezing by allowing vapor to pass through the holes in the side of the inner tube. The 55/50 standard taper joint connects the trap body with the stopper. Hose connections are 15 mm OD at the largest serration.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty