Would you like to log in again?
Dangerous Goods UN 4GV Packaging, Qorpak
Supplier: Qorpak
HMS-6600M
14002-616CS 822.76 USD InStock
14002-616
Dangerous Goods UN 4GV Packaging, Qorpak
Mailers and Shippers
The 4GV Slip Cover Can Packaging offers maximum protection for 1 liter or smaller bottles. Ideal for regulated transport of corrosives and materials requiring a secondary metal package.
  • Tested with vermiculite and polypropylene absorbent
The 4GV marking allows for the use of alternate primary containers, given the packing instructions are followed. For air shipments, a pressure tested primary container or pressure tested bag around the primary must be used.
Certifications: Meets UN 4GV standards for Packing Groups I, II, and III.
Ordering information: Absorbent and bottle not included.
Packaging: Sold in a convenience pack of 12.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty