Would you like to log in again?
Dangerous Goods Four Bottle UN 4G Packaging, Qorpak
Supplier: Qorpak
HMS-6466M
16151-706CS 170.69 USD InStock
16151-706
Dangerous Goods Four Bottle UN 4G Packaging, Qorpak
Mailers and Shippers
Maximize the number of bottles shipped with multiple bottle packaging. Compliance is made easy by adding absorbent sheets and a bag to the package.
Variation 1 of 49 CFR 178.601(g)(1) allows for some flexibility of primary containers, given the packing instructions are followed. For air shipments, a pressure tested primary container or pressure tested bag around the primary must be used.
Certifications: Meets UN 4G standards for Packing Groups II and III.
Ordering information: Includes polypropylene absorbent and bag. Bottles not included.
Packaging: Sold in a convenience pack of 4.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty