Would you like to log in again?
Stopcock Adapter with Hose Connection, Ace Glass Incorporated
Supplier: Ace Glass
5202-116
76214-388EA 151.14 USD InStock
76214-388
Stopcock Adapter with Hose Connection, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Stopcock adapter featuring a straight hose connection and a PTFE plug.
  • High-quality borosilicate glass
  • Features 45/50 standard taper joint
Delivery information: Includes 1:5 PTFE straight bore plug.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty