Would you like to log in again?
Versaflo™ Prefilter™
Supplier: 3M Healthcare
Versaflo™
7100021255
10152-902CS 469.2 USD InStock
10152-902
Versaflo™ Prefilter™
Filters Gas Filters
Versaflo™ Prefilter helps in reducing the operating costs by extending the life of TR-6000 series filters and cartridges.
  • Easy to use due to the simple maintenance-free design
  • Flexible application, as TR-6600 may be used with TR-662 spark arrestor
The TR-6600 Prefilter can extend the service life of the particulate (HE) element of TR-600 series filters and cartridges by capturing large particles that may otherwise clog the HE filter. Frequent prefilter changeout may help optimize PAPR system performance. The TR-6600 prefilter must be installed and used according to the User Instructions. Use of the TR-6600 may reduce PAPR operating costs by extending the service life of the main filter/cartridge. The TR-6600 is maintenance-free and should be discarded when loaded or damaged.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty