Would you like to log in again?
NFκB p65 (Total/Phospho) ELISA kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-EKS-446A
76218-676EA 878.51 USD InStock
76218-676
NFκB p65 (Total/Phospho) ELISA kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of active NFκB p65 in cell lysates of human, mouse, and rat origin.
The NFκB p65 EIA kit provides a sensitive, rapid and reliable method to measure the active form of NFκB p65. The assay uses streptavidin-coated plates with bound NFκB biotinylated-consensus sequence to capture only the active form of NFκB. The captured active NFκB is incubated with a specific NFκB p65 antibody, which is then detected using an HRP conjugated secondary antibody. The assay is developed with a chemiluminescent substrate and the signal is detected using a luminometer. Higher throughput 96 well plate based method faster than EMSA and Western blot with sensitive chemiluminescent detection.
  • Fast- measure the active form of NFκB p65 in just 1.5 hours
  • Higher throughput - 96 well plate based method faster than EMSA and Western blot
  • Sensitive - colorimetric detection
Ordering information: Contains Microtiter plate, cell lysis buffer, 10X wash buffer, detection reagent, stop solution, plate sealer, capture antibody, detector antibody, positive control lysate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty