Would you like to log in again?
Versaflo™ Spark Arrestor, 3M™
Supplier: 3M
Versaflo™
7100035845
10152-904CS 18.21 USD InStock
10152-904
Versaflo™ Spark Arrestor, 3M™
Welding Supplies
Protects TR-6000 series filters and cartridges from potential damage due to sparks.
  • May reduce operating costs by extending the life of TR-6000 series filters and cartridges
  • May lower operating costs when used as a cleanable pre-filter
  • Flexible application, as TR-662 may be used with TR-6600 prefilter
The TR-662 spark arrestor may be used with all TR-600 series filters and cartridges. It helps prevent sparks and other hot particles from damaging the filter/cartridge and must be used whenever sparks or hot airborne particles are present in the workplace. The TR-662 spark arrestor also acts as a pre-filter by capturing many of the larger particles that would otherwise be drawn into and load the main filter. The TR-662 may be cleaned and reused in accordance with the User Instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty