Would you like to log in again?
Oakton PC 2700 Benchtop pH/Conductivity Meters, Cole-Parmer
Supplier: Cole Parmer
Oakton®
WD-35414-00 35414-20 WD-35414-00
101076-386EA 2578.61 USD InStock
101076-386 76221-206 76221-208
Oakton PC 2700 Benchtop pH/Conductivity Meters, Cole-Parmer
Electrochemical Meters pH and Multi-Parameter Meters
Multiparameter benchtop meters are an all-in-one solution for laboratory research work. These meters measure pH, mV, conductivity, TDS, and temperature—ideal for water quality testing in the lab.
  • Advanced research-grade meter
  • Includes Ion concentration readings
  • Features pH auto-buffer recognition for both USA and NIST buffer sets
  • Automatic or manual temperature compensation
  • Memory storage for quick and easy retrieval
Meters offer an advanced user interface and advanced measurement capabilities. Meters meet GLP requirements meters stamp all calibration and stored data with time/date. Nonvolatile memory stores up to 500 data sets to simplify documenting large volumes of pH, mV, ISE, conductivity, TDS, salinity, or resistivity measurements. Data is easily exported to a computer via RS232 port.
Ordering information: Includes universal 100/240 V AC power supply and electrode stand. Meter with probes adds combination pH electrode and conductivity/temperature probe. RS232 port needs to be ordered seperately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty