Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium-tert-butanolate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: Lithium tert-butoxide

03-0780-25G 03-0780-100G
100209-286EA 396.31 USD InStock
100209-286 100199-686
Lithium-tert-butanolate ≥98%
Lithium tert-butanolate
Alkoxide, Volatile Precursors for CVD


Formula: C₄H₉LiO
Density: 0.89
MDL Number: MFCD00050479
CAS Number: 1907-33-1
UN: 3206
ADR: 4.2,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty