Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Decaborane ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

05-0500-1G 05-0500-250MG
100210-720EA 346.77 USD InStock
100210-720 100206-190
Decaborane ≥97%
Decaborane


Formula: B₁₀H₁₄
Boiling Pt: 213 °C
Melting Pt: 100 °C
Density: 0.94
MDL Number: MFCD00011314
CAS Number: 17702-41-9
UN: 1868
ADR: 4.1,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty