Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium-2-ethylhexanoate hydrate ≥95%

Supplier: STREM CHEMICALS INC MS
Warning
Synonyms: Potassium 2-ethylhexanoate hydrate

19-1920-25G 19-1920-5G
100208-846EA 90.58 USD InStock
100208-846 100208-844
Potassium-2-ethylhexanoate hydrate ≥95%
Potassium-2-ethylhexanoate hydrate


Formula: C₈H₁₅KO₂·xH₂O
MW: 182.30 g/mol (anhy)
MDL Number: MFCD00149471
CAS Number: 3164-85-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty