Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vanadium(IV) oxide meso-tetraphenylporphine

Supplier: STREM CHEMICALS INC MS
Warning
Synonyms: Vanadyl meso-tetraphenylporphine

23-6000-2G 23-6000-500MG
100210-622EA 270.34 USD InStock
100210-622 100197-014
Vanadium(IV) oxide meso-tetraphenylporphine
Vanadium(IV) oxide meso-tetraphenylporphine
Porphine


Formula: C₄₄H₂₈N₄OV
MW: 679.66 g/mol
MDL Number: MFCD00012154
CAS Number: 14705-63-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty