Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dicarbonylcyclopentadienylcobalt ≥95%

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: Cyclopentadienylcobalt dicarbonylDicarbonylcyclopentadienylcobalt(I)

27-0550-10G 27-0550-2G
100207-634EA 281.66 USD InStock
100207-634 100207-636
Dicarbonylcyclopentadienylcobalt ≥95%
Dicarbonylcyclopentadienylcobalt
Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD


Formula: C₇H₅CoO₂
MW: 180.05 g/mol
Boiling Pt: 37…38.5 °C (2 mmHg)
Density: 1,35
Flash Pt: 80 °F
MDL Number: MFCD00009589
CAS Number: 12078-25-0
UN: 1993
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty