Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dodecacarbonyltetracobalt ≥98%

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: Tetracobalt dodecacarbonyl

27-1950-5G 27-1950-25G 27-1950-1G
100210-634EA 458.58 USD InStock
100210-634 100210-632 100205-062
Dodecacarbonyltetracobalt ≥98%
Dodecacarbonyltetracobalt
Metal Carbonyls


Formula: C₁₂Co₄O₁₂
MW: 571.853 g/mol
Melting Pt: 60 °C (dec.)
Density: 2.09
UN: 3178
ADR: 4.1,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty