Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Yttrium(III) isopropoxide 20 - 25% in toluene

Supplier: STREM CHEMICALS INC MS
Danger

39-3000-5G 39-3000-25G 39-3000-100G
100201-210EA 2549.11 USD InStock
100201-210 100203-772 100198-178
Yttrium(III) isopropoxide 20 - 25% in toluene
Yttrium(III) isopropoxide
Alkoxide


Formula: C₉H₂₁O₃Y
MW: 266.17 g/mol
Flash Pt: 40 °F
MDL Number: MFCD00015641
CAS Number: 2172-12-5
UN: 1993
ADR: 3,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty