Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclopentadienylniobium(V) tetrachloride ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

41-0500-5G 41-0500-1G
100203-130EA 141.54 USD InStock
100203-130 100199-882
Cyclopentadienylniobium(V) tetrachloride ≥98%
Cyclopentadienylniobium(V) tetrachloride
Metallocenes, Derivatives & Cp Precursors


Formula: C₅H₅Cl₄Nb
Melting Pt: 180 °C (dec.)
MDL Number: MFCD00070445
CAS Number: 33114-15-7
UN: 1759
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty