Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(cyclopentadienyl)ruthenium ≥99% (99.9%-Ru)

Supplier: STREM CHEMICALS INC MS
Warning
Synonyms: Bis(cyclopentadienyl)ruthenium(II)Ruthenocene

44-6200-5G 44-6200-1G
100211-100EA 176.92 USD InStock
100211-100 100211-102
Bis(cyclopentadienyl)ruthenium ≥99% (99.9%-Ru)
Bis(cyclopentadienyl)ruthenium
Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD


Formula: C₁₀Ru
MW: 221.18 g/mol
Melting Pt: 194…198 °C
MDL Number: MFCD00001442
CAS Number: 1287-13-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty