Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Samarium(III) carbonate hydrate ≥99.9% (Sm basis)

Supplier: STREM CHEMICALS INC MS

62-2000-10G 62-2000-50G
100203-246EA 602.95 USD InStock
100203-246 100205-182
Samarium(III) carbonate hydrate ≥99.9% (Sm basis)
Samarium(III) carbonate hydrate


Formula: Sm₂(CO₃)₃·xH₂O
MW: 480.73 g/mol (anhy)
Melting Pt: >500 °C (dec.)
MDL Number: MFCD00150444
CAS Number: 5895-47-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty