Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gadolinium(III) carbonate hydrate ≥99.99% (Gd basis), PURATREM

Supplier: STREM CHEMICALS INC MS

64-3000-50G 64-3000-10G
100210-032EA 472.74 USD InStock
100210-032 100210-030
Gadolinium(III) carbonate hydrate ≥99.99% (Gd basis), PURATREM
Gadolinium(III) carbonate hydrate


Formula: Gd₂(CO₃)₃.xH₂O
MDL Number: MFCD00150240
CAS Number: 38245-36-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty