Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Osmium tetraoxide (∼10% OsO₄) on polystyrene, microencapsulated

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: Osmic acid anhydrideOsmic acidOsmium (VIII) oxide

76-2956-1G
100207-742EA 1116.74 USD InStock
100207-742
Osmium tetraoxide (∼10% OsO₄) on polystyrene, microencapsulated
Osmium tetraoxide


Formula: O₄Os
MW: 254.23 g/mol
MDL Number: MFCD00011150
CAS Number: 20816-12-0
UN: 2471
ADR: 6.1,I
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty