Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium trichloroammineplatinate(II)

Supplier: STREM CHEMICALS INC MS
Synonyms: Potassium amminetrichloroplatinate(II)

78-1975-500MG
100210-668EA 918.58 USD InStock
100210-668
Potassium trichloroammineplatinate(II)
Potassium trichloroammineplatinate(II)


Formula: K(PtCl₃NH₃)
MW: 357.57 g/mol
MDL Number: MFCD00145154
CAS Number: 13820-91-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty