Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium dicyanoaurate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: Potassium dicyanoaurate(I)Gold(I) potassium cyanide

79-3000-5G 79-3000-1G
100196-400EA 859.14 USD InStock
100196-400 100196-398
Potassium dicyanoaurate ≥99%
Potassium dicyanoaurate


Formula: C₂AuKN₂
MW: 288.1 g/mol
MDL Number: MFCD00011414
CAS Number: 13967-50-5
UN: 1588
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty