Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium methoxide 10% in methanol

Supplier: STREM CHEMICALS INC MS
Danger

93-0335-500G 93-0335-100G
100205-236EA 62.28 USD InStock
100205-236 100199-466
Lithium methoxide 10% in methanol
Lithium methoxide
Alkoxide


Formula: CH₃LiO
MW: 37.98 g/mol
Flash Pt: 52 °F
MDL Number: MFCD00036357
CAS Number: 865-34-9
UN: 1992
ADR: 3,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty